Assay Details
Target Gene Details
Entrez Gene ID: | 21849 |
Gene Name: | tripartite motif-containing 28 |
Gene Aliases: |
AA408787, KAP-1, KRIP-1, MommeD9, Tif1b, Tif1beta |
Location: |
Chr.7:13024152-13031032 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim28 | NM_011588.3 | NP_035718.2 | ||
| AF230391.1 | AAG50170.1 | |||
| AF230392.1 | AAG50171.1 | |||
| AK141357.1 | BAE24661.1 | |||
| AK179925.1 | ||||
| AK184928.1 | ||||
| AK217247.1 | ||||
| BC058391.1 | AAH58391.1 | |||
| BC089337.1 | AAH89337.1 | |||
| U67303.1 | AAB17272.1 | |||
| X99644.1 | CAA67963.1 |
More Information
Set Membership: |
|

Genomic Map