Assay Details
Target Gene Details
Entrez Gene ID: | 19655 |
Gene Name: | RNA binding motif protein, X chromosome |
Gene Aliases: |
hnRNP G |
Location: |
Chr.X:57383348-57393042 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbmx | NM_001166623.1 | NP_001160095.1 | ||
| NM_011252.4 | NP_035382.1 | |||
| NR_029425.1 | ||||
| AJ237846.1 | CAB51361.1 | |||
| AJ237847.1 | CAB51362.1 | |||
| AK083899.1 | ||||
| AK135164.1 | BAE22448.1 | |||
| AK165006.1 | BAE37998.1 | |||
| AK180208.1 | ||||
| BC003710.1 | AAH03710.1 | |||
| CK619886.1 |
More Information
Set Membership: |
|

Genomic Map