Assay Details
Target Gene Details
Entrez Gene ID: | 50506 |
Gene Name: | dual oxidase 2 |
Gene Aliases: |
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2 |
Location: |
Chr.15:45092653-45114161 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUOX2 | NM_014080.4 | 18 | 2454 | NP_054799.4 |
| XM_005254421.2 | 18 | 2454 | XP_005254478.1 | |
| AB209010.1 | 12 | 3982 | BAD92247.1 | |
| AF181972.1 | 10 | 1237 | AAF20055.1 | |
| AF230496.1 | 18 | 2436 | AAF73922.1 | |
| AF267981.1 | 18 | 2454 | AAF78954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map