Assay Details
Target Gene Details
Entrez Gene ID: | 414152 |
Gene Name: | chromosome 10 open reading frame 105 |
Gene Aliases: |
- |
Location: |
Chr.10:71711701-71737824 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf105 | NM_001168390.1 | NP_001161862.1 | ||
| AK074172.1 |
Target Gene Details
Entrez Gene ID: | 64072 |
Gene Name: | cadherin-related 23 |
Gene Aliases: |
CDHR23, USH1D |
Location: |
Chr.10:71396934-71815947 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 36 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470963 | Chr.10:71687524 - 71749708 on Build GRCh38 | Loss |
|
| nsv831910 | Chr.10:71635558 - 71843223 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72817951] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map