Assay Details
Target Gene Details
Entrez Gene ID: | 9940 |
Gene Name: | deleted in lung and esophageal cancer 1 |
Gene Aliases: |
CFAP81, DLC-1, DLC1, F56 |
Location: |
Chr.3:38038595-38122741 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEC1 | NM_001321153.1 | 1 | 143 | NP_001308082.1 |
| NM_007335.3 | 1 | 143 | NP_031361.2 | |
| NM_007337.3 | 1 | 143 | NP_031363.2 | |
| XM_006713438.3 | 1 | 752 | XP_006713501.1 | |
| XM_006713439.3 | 1 | 751 | XP_006713502.1 | |
| XM_006713440.3 | 1 | 752 | XP_006713503.1 | |
| XM_011534317.2 | 1 | 751 | XP_011532619.1 | |
| XM_011534318.2 | 1 | 752 | XP_011532620.1 | |
| AB020522.1 | 1 | 143 | BAA77247.1 | |
| AB425198.1 | 1 | 115 | ||
| BC129824.1 | 1 | 143 | ||
| BC129825.1 | 1 | 143 | ||
| BC171799.1 | 1 | 143 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819131 | Chr.3:38037529 - 38044812 on Build GRCh38 | Loss |
|
| esv2759140 | Chr.3:37789992 - 38062733 on Build GRCh38 | Loss |
|
| nsv590101 | Chr.3:38000691 - 38065526 on Build GRCh38 | Loss |
|
| nsv428416 | Chr.3:37864474 - 38062733 on Build GRCh38 | Loss |
|
| nsv508911 | Chr.3:38019295 - 38091072 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map