Assay Details
Target Gene Details
Entrez Gene ID: | 6337 |
Gene Name: | sodium channel epithelial 1 alpha subunit |
Gene Aliases: |
BESC2, ENaCa, ENaCalpha, SCNEA, SCNN1 |
Location: |
Chr.12:6346843-6377357 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCNN1A | NM_001038.5 | 13 | 2849 | NP_001029.1 |
| NM_001159575.1 | 13 | 2720 | NP_001153047.1 | |
| NM_001159576.1 | 12 | 3001 | NP_001153048.1 | |
| AK025536.1 | 8 | 1573 | ||
| AK172792.1 | 12 | 3004 | ||
| BC006526.2 | 12 | 2687 | AAH06526.2 | |
| BC062613.1 | 12 | 2616 | AAH62613.1 | |
| BU680600.1 | ||||
| X76180.1 | 13 | 2659 | CAA53773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745469 | Chr.12:6346988 - 6347463 on Build GRCh38 | Deletion |
|
| esv34325 | Chr.12:6342424 - 6513183 on Build GRCh38 | Gain |
|
| esv2759876 | Chr.12:6344718 - 6554669 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map