Assay Details
Target Gene Details
Entrez Gene ID: | 1967 |
Gene Name: | eukaryotic translation initiation factor 2B subunit alpha |
Gene Aliases: |
EIF2B, EIF2BA |
Location: |
Chr.12:123621023-123633776 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2B1 | NM_001414.3 | NP_001405.1 | ||
| AL541292.3 | ||||
| BC103763.1 | AAI03764.1 | |||
| BC104188.1 | AAI04189.1 | |||
| BC104189.2 | AAI04190.1 | |||
| CR456831.1 | CAG33112.1 | |||
| X95648.1 | CAA64950.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1566n100 | Chr.12:123610211 - 123711309 on Build GRCh38 | Gain |
|
| dgv524e212 | Chr.12:123614065 - 123625575 on Build GRCh38 | Loss |
|
| nsv1042181 | Chr.12:123552853 - 123668403 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map