Assay Details
Target Gene Details
Entrez Gene ID: | 3784 |
Gene Name: | potassium voltage-gated channel subfamily Q member 1 |
Gene Aliases: |
ATFB1, ATFB3, JLNS1, KCNA8, KCNA9, KVLQT1, Kv1.9, Kv7.1, LQT, LQT1, RWS, SQT2, WRS |
Location: |
Chr.11:2444991-2849110 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ1 | NM_000218.2 | NP_000209.2 | ||
| NM_181798.1 | 1 | 124 | NP_861463.1 | |
| AF000571.1 | AAC51776.1 | |||
| AF003743.1 | AAC00066.1 | |||
| AF051426.1 | 1 | 124 | AAC05705.1 | |
| AI439544.1 | ||||
| AJ006344.1 | 1 | 124 | ||
| AK129678.1 | 1 | 122 | ||
| AK290618.1 | 1 | 115 | ||
| AY114213.1 | AAM94040.1 | |||
| EF010935.1 | 1 | 142 | ||
| U89364.1 | AAC51781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951287 | Chr.11:2396771 - 2475670 on Build GRCh38 | Deletion |
|
| nsv469928 | Chr.11:2294174 - 2473462 on Build GRCh38 | Loss |
|
| nsv1037537 | Chr.11:2409367 - 2810311 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map