Assay Details
Target Gene Details
Entrez Gene ID: | 1649 |
Gene Name: | DNA damage inducible transcript 3 |
Gene Aliases: |
CEBPZ, CHOP, CHOP-10, CHOP10, GADD153 |
Location: |
Chr.12:57516588-57520517 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDIT3 | NM_001195053.1 | 4 | 692 | NP_001181982.1 |
| NM_001195054.1 | 4 | 639 | NP_001181983.1 | |
| NM_001195055.1 | 4 | 616 | NP_001181984.1 | |
| NM_001195056.1 | 3 | 802 | NP_001181985.1 | |
| NM_001195057.1 | 4 | 521 | NP_001181986.1 | |
| NM_004083.5 | 4 | 535 | NP_004074.2 | |
| AA476561.1 | 4 | 542 | ||
| AK316581.1 | 3 | 796 | ||
| AV729744.1 | 4 | 521 | ||
| AY461580.1 | 4 | 694 | ||
| BC003637.2 | 4 | 536 | AAH03637.1 | |
| BM464290.1 | 4 | 525 | ||
| BT006691.1 | 2 | 355 | AAP35337.1 | |
| BU664077.1 | ||||
| CN482428.1 | 4 | 499 | ||
| S40706.1 | 4 | 526 | AAB22646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map