Assay Details
Target Gene Details
Entrez Gene ID: | 226 |
Gene Name: | aldolase, fructose-bisphosphate A |
Gene Aliases: |
ALDA, GSD12, HEL-S-87p |
Location: |
Chr.16:30053090-30070420 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDOA | NM_000034.3 | 5 | 542 | NP_000025.1 |
| XM_011545768.2 | 2 | 405 | XP_011544070.1 | |
| AK098778.1 | 3 | 583 | ||
| AK308495.1 | 5 | 461 | ||
| AK308748.1 | 4 | 354 | ||
| BC004333.1 | 5 | 462 | AAH04333.1 | |
| DB057572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833187 | Chr.16:30051939 - 30173160 on Build GRCh38 | Loss |
|
| nsv1160384 | Chr.16:30023148 - 30178660 on Build GRCh38 | Duplication |
|
| esv2758642 | Chr.16:30062634 - 30371959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map