Assay Details
Target Gene Details
Entrez Gene ID: | 414152 |
Gene Name: | chromosome 10 open reading frame 105 |
Gene Aliases: |
- |
Location: |
Chr.10:71711701-71737824 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf105 | NM_001168390.1 | NP_001161862.1 | ||
| AK074172.1 |
Target Gene Details
Entrez Gene ID: | 64072 |
Gene Name: | cadherin-related 23 |
Gene Aliases: |
CDHR23, USH1D |
Location: |
Chr.10:71396934-71815947 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH23 | NM_001171930.1 | 32 | 4365 | NP_001165401.1 |
| NM_022124.5 | 32 | 4365 | NP_071407.4 | |
| XM_006717940.3 | 33 | 4419 | XP_006718003.1 | |
| XM_006717942.3 | 31 | 4353 | XP_006718005.1 | |
| XM_011540039.2 | 33 | 4419 | XP_011538341.1 | |
| XM_011540042.2 | 33 | 4419 | XP_011538344.1 | |
| XM_011540043.2 | 33 | 4419 | XP_011538345.1 | |
| XM_011540044.2 | 33 | 4060 | XP_011538346.1 | |
| XM_011540045.2 | 33 | 4419 | XP_011538347.1 | |
| XM_011540046.2 | 28 | 3854 | XP_011538348.1 | |
| XM_011540047.2 | 22 | 3154 | XP_011538349.1 | |
| XM_011540048.2 | 33 | 4419 | XP_011538350.1 | |
| XM_011540049.2 | 33 | 4419 | XP_011538351.1 | |
| XM_011540051.2 | 33 | 4419 | XP_011538353.1 | |
| XM_011540052.2 | 5 | 694 | XP_011538354.1 | |
| XM_017016499.1 | 32 | 4365 | XP_016871988.1 | |
| XM_017016500.1 | 33 | 4419 | XP_016871989.1 | |
| XM_017016501.1 | 12 | 1921 | XP_016871990.1 | |
| XM_017016502.1 | 12 | 2109 | XP_016871991.1 | |
| XM_017016503.1 | XP_016871992.1 | |||
| AF312024.1 | 31 | 3975 | AAG48303.1 | |
| AK093818.1 | 4 | 667 | BAC04231.1 | |
| AY010111.2 | 31 | 3975 | AAG27034.2 | |
| BC065284.1 | 32 | 4342 | AAH65284.1 | |
| DB333699.1 | 1 | 479 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470963 | Chr.10:71687524 - 71749708 on Build GRCh38 | Loss |
|
| nsv831910 | Chr.10:71635558 - 71843223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map