Assay Details
Target Gene Details
Entrez Gene ID: | 2581 |
Gene Name: | galactosylceramidase |
Gene Aliases: |
- |
Location: |
Chr.14:87933014-87993665 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALC | NM_000153.3 | 7 | 815 | NP_000144.2 |
| NM_001201401.1 | 6 | 746 | NP_001188330.1 | |
| NM_001201402.1 | 7 | 796 | NP_001188331.1 | |
| XM_011536618.2 | 7 | 1097 | XP_011534920.1 | |
| AK294040.1 | 8 | 660 | ||
| AK296530.1 | 6 | 637 | ||
| AK302519.1 | 2 | 231 | ||
| AK302683.1 | 7 | 969 | ||
| AK302956.1 | 7 | 635 | ||
| BC036518.2 | 7 | 667 | AAH36518.1 | |
| BU675449.1 | 4 | 540 | ||
| D25283.1 | 7 | 698 | BAA04971.1 | |
| D25284.1 | 7 | 698 | BAA04972.1 | |
| L23116.1 | 7 | 707 | AAA16645.1 |
More Information
Set Membership: |
|

Genomic Map