Assay Details
Target Gene Details
Entrez Gene ID: | 5194 |
Gene Name: | peroxisomal biogenesis factor 13 |
Gene Aliases: |
NALD, PBD11A, PBD11B, ZWS |
Location: |
Chr.2:61017677-61051990 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEX13 | NM_002618.3 | 4 | 1178 | NP_002609.1 |
| AB022192.1 | 4 | 1133 | BAA88907.1 | |
| AF048755.1 | 4 | 1133 | AAC39844.1 | |
| AK315244.1 | 4 | 1137 | ||
| BC067090.1 | 4 | 1120 | AAH67090.1 | |
| U71374.1 | 4 | 1095 | AAD05572.1 |
More Information
Set Membership: |
|

Genomic Map