Assay Details
Target Gene Details
Entrez Gene ID: | 1439 |
Gene Name: | colony stimulating factor 2 receptor beta common subunit |
Gene Aliases: |
CD131, CDw131, IL3RB, IL5RB, SMDP5 |
Location: |
Chr.22:36913574-36940449 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSF2RB | NM_000395.2 | NP_000386.1 | ||
| XM_005261340.3 | XP_005261397.1 | |||
| XM_011529903.2 | XP_011528205.1 | |||
| XM_011529904.2 | XP_011528206.1 | |||
| XM_011529905.2 | XP_011528207.1 | |||
| AK302993.1 | ||||
| BC070085.1 | AAH70085.1 | |||
| CR456428.1 | CAG30314.1 | |||
| M59941.1 | AAA18171.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758837 | Chr.22:36779110 - 36964864 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map