Assay Details
Target Gene Details
Entrez Gene ID: | 4893 |
Gene Name: | neuroblastoma RAS viral oncogene homolog |
Gene Aliases: |
ALPS4, CMNS, N-ras, NCMS, NRAS1, NS6 |
Location: |
Chr.1:114704464-114716894 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRAS | NM_002524.4 | NP_002515.1 | ||
| AF493919.1 | AAM12633.1 | |||
| BC005219.1 | AAH05219.1 | |||
| BT019734.1 | AAV38539.1 | |||
| BT019735.1 | AAV38540.1 | |||
| DA007844.1 | ||||
| X02751.1 | CAA26529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map