Assay Details
Target Gene Details
Entrez Gene ID: | 2261 |
Gene Name: | fibroblast growth factor receptor 3 |
Gene Aliases: |
ACH, CD333, CEK2, HSFGFR3EX, JTK4 |
Location: |
Chr.4:1793299-1808872 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGFR3 | NM_000142.4 | NP_000133.1 | ||
| NM_001163213.1 | NP_001156685.1 | |||
| NM_022965.3 | NP_075254.1 | |||
| XM_006713868.1 | XP_006713931.1 | |||
| XM_006713869.1 | XP_006713932.1 | |||
| XM_006713870.1 | XP_006713933.1 | |||
| XM_006713871.1 | XP_006713934.1 | |||
| XM_006713872.1 | XP_006713935.1 | |||
| XM_006713873.1 | XP_006713936.1 | |||
| XM_011513420.1 | XP_011511722.1 | |||
| XM_011513422.1 | XP_011511724.1 | |||
| AB209441.1 | BAD92678.1 | |||
| AF238374.1 | AAF97749.1 | |||
| AF245114.1 | AAF63380.1 | |||
| AK308936.1 | ||||
| BC121175.2 | ||||
| BC128610.1 | ||||
| BC153824.1 | ||||
| HM244683.1 | ||||
| KJ534834.1 | ||||
| KJ534974.1 | ||||
| KJ535004.1 | ||||
| M58051.1 | AAA52450.1 | |||
| M64347.1 | AAA58470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8902n54 | Chr.4:1797575 - 1807760 on Build GRCh38 | Loss |
|
| nsv428434 | Chr.4:1623665 - 1813048 on Build GRCh38 | Gain |
|
| nsv949756 | Chr.4:1710274 - 1817773 on Build GRCh38 | Deletion |
|
| dgv8901n54 | Chr.4:1644637 - 1864631 on Build GRCh38 | Loss |
|
| nsv508989 | Chr.4:1623823 - 1846590 on Build GRCh38 | Insertion |
|
| nsv593339 | Chr.4:1803751 - 1807468 on Build GRCh38 | Loss |
|
| nsv829836 | Chr.4:1775678 - 1901525 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map