Assay Details
Target Gene Details
Entrez Gene ID: | 2165 |
Gene Name: | coagulation factor XIII B chain |
Gene Aliases: |
FXIIIB |
Location: |
Chr.1:197038741-197067267 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F13B | NM_001994.2 | NP_001985.2 | ||
| XM_011509283.2 | XP_011507585.1 | |||
| XM_011509284.2 | XP_011507586.1 | |||
| XM_011509286.2 | XP_011507588.1 | |||
| AK290560.1 | ||||
| M14057.1 | AAA88042.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3588491 | Chr.1:197008580 - 197058939 on Build GRCh38 | Gain |
|
| esv2758990 | Chr.1:196621707 - 197200642 on Build GRCh38 | Gain+Loss |
|
| nsv528460 | Chr.1:196573437 - 197477572 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map