Assay Details
Target Gene Details
Entrez Gene ID: | 102465841 |
Gene Name: | microRNA 7855 |
Gene Aliases: |
hsa-mir-7855 |
Location: |
Chr.14:64785626-64785686 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR7855 | NR_107009.1 |
Target Gene Details
Entrez Gene ID: | 6710 |
Gene Name: | spectrin beta, erythrocytic |
Gene Aliases: |
EL3, HS2, HSPTB1, SPH2 |
Location: |
Chr.14:64746283-64879892 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTB | NM_000347.5 | NP_000338.3 | ||
| NM_001024858.2 | NP_001020029.1 | |||
| XM_005268023.4 | XP_005268080.1 | |||
| XM_011537105.2 | XP_011535407.1 | |||
| XM_017021612.1 | XP_016877101.1 | |||
| XM_017021613.1 | XP_016877102.1 | |||
| XM_017021614.1 | XP_016877103.1 | |||
| AB209415.1 | BAD92652.1 | |||
| BC136284.1 | ||||
| BC136285.1 | ||||
| J05500.1 | AAA60578.1 | |||
| M18054.1 | AAA60572.1 | |||
| M37884.1 | AAA63259.1 | |||
| M57948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map