Assay Details
Target Gene Details
Entrez Gene ID: | 57167 |
Gene Name: | spalt like transcription factor 4 |
Gene Aliases: |
DRRS, HSAL4, ZNF797 |
Location: |
Chr.20:51782717-51802523 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SALL4 | NM_001318031.1 | 4 | 1685 | NP_001304960.1 |
| NM_020436.4 | 4 | 2996 | NP_065169.1 | |
| XM_005260467.3 | 4 | 2977 | XP_005260524.1 | |
| XM_011528921.2 | 6 | 3172 | XP_011527223.1 | |
| XM_011528922.2 | 5 | 3131 | XP_011527224.1 | |
| AA419623.1 | ||||
| AI638036.1 | 1 | 506 | ||
| AK001666.1 | 4 | 2946 | ||
| AY170621.1 | 4 | 1570 | AAO16566.1 | |
| AY170622.1 | 3 | 550 | AAO16567.1 | |
| AY172738.1 | 4 | 2881 | AAO44950.1 | |
| BC111714.1 | 4 | 2937 | AAI11715.1 | |
| DA747867.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758800 | Chr.20:51706078 - 51802856 on Build GRCh38 | Loss |
|
| nsv964409 | Chr.20:51782238 - 51784662 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map