Assay Details
Target Gene Details
Entrez Gene ID: | 26010 |
Gene Name: | spermatogenesis associated serine rich 2 like |
Gene Aliases: |
DNAPTP6, SGNP |
Location: |
Chr.2:200305881-200482263 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATS2L | NM_001282735.1 | 1 | 234 | NP_001269664.1 |
| NM_001282743.1 | 1 | 234 | NP_001269672.1 | |
| NM_015535.2 | 1 | 234 | NP_056350.2 | |
| AK090867.1 | ||||
| AK290496.1 | 1 | 222 | ||
| AL110124.1 | 1 | 42 | CAB53658.2 | |
| AY450394.1 | 1 | 42 | AAR21087.1 | |
| BP310992.1 | 1 | 234 | ||
| CN480518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3107 | Chr.2:200270658 - 200315985 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map