Assay Details
Target Gene Details
Entrez Gene ID: | 8490 |
Gene Name: | regulator of G-protein signaling 5 |
Gene Aliases: |
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129 |
Location: |
Chr.1:163142299-163321791 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS5 | NM_001195303.2 | 3 | 5215 | NP_001182232.1 |
| NM_001254748.1 | 6 | 5273 | NP_001241677.1 | |
| NM_001254749.1 | 5 | 5400 | NP_001241678.1 | |
| NM_003617.3 | 5 | 5388 | NP_003608.1 | |
| AF113212.1 | 1 | 1356 | AAG39283.1 | |
| AF172821.1 | 1 | 1479 | AAQ13615.1 | |
| AF173551.1 | 1 | 1773 | AAQ13619.1 | |
| AF176919.1 | 1 | 1356 | AAQ13663.1 | |
| AK026775.1 | 1 | 1312 | ||
| AL157502.1 | 1 | 554 | ||
| BX537427.1 | 5 | 5132 | CAD97669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890704 | Chr.1:163103712 - 163233339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map