Assay Details
Target Gene Details
Entrez Gene ID: | 112849 |
Gene Name: | trans-L-3-hydroxyproline dehydratase |
Gene Aliases: |
C14orf149 |
Location: |
Chr.14:59465545-59484438 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L3HYPDH | NM_144581.1 | 2 | 597 | NP_653182.1 |
| XM_005267298.3 | 2 | 234 | XP_005267355.1 | |
| XM_011536386.2 | 2 | 680 | XP_011534688.1 | |
| XM_011536387.1 | 2 | 680 | XP_011534689.1 | |
| XM_011536388.1 | 2 | 680 | XP_011534690.1 | |
| AK058165.1 | 2 | 597 | BAB71696.1 | |
| AK294843.1 | 2 | 779 | ||
| BC012131.1 | 2 | 610 | AAH12131.1 |
More Information
Set Membership: |
|

Genomic Map