Assay Details
Target Gene Details
Entrez Gene ID: | 11331 |
Gene Name: | prohibitin 2 |
Gene Aliases: |
BAP, BCAP37, Bap37, PNAS-141, REA, hBAP, p22 |
Location: |
Chr.12:6965352-6970823 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHB2 | NM_001144831.1 | NP_001138303.1 | ||
| NM_001267700.1 | NP_001254629.1 | |||
| AF126021.1 | AAF17231.1 | |||
| AF150962.1 | AAD38042.1 | |||
| AF178980.1 | AAF44345.1 | |||
| AK298217.1 | ||||
| AK301316.1 | ||||
| BC014766.1 | AAH14766.1 | |||
| BC110322.1 | AAI10323.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain |
|
| nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion |
|
| nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map