Assay Details
Target Gene Details
Entrez Gene ID: | 10237 |
Gene Name: | solute carrier family 35 member B1 |
Gene Aliases: |
UGTREL1 |
Location: |
Chr.17:49700934-49708173 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35B1 | NM_001278784.1 | NP_001265713.1 | ||
| NM_005827.2 | NP_005818.2 | |||
| XM_005256929.3 | XP_005256986.1 | |||
| XM_006721632.3 | XP_006721695.1 | |||
| XM_011524179.2 | XP_011522481.1 | |||
| XM_011524180.2 | XP_011522482.1 | |||
| AK098056.1 | ||||
| AK124975.1 | 2 | 922 | ||
| AK293563.1 | ||||
| BC011888.2 | AAH11888.1 | |||
| BM476321.1 | ||||
| BX641166.1 | CAE46073.1 | |||
| CR745911.1 | ||||
| CX164572.1 | ||||
| D87989.1 | BAA13525.1 |
More Information
Set Membership: |
|

Genomic Map