Assay Details
Target Gene Details
Entrez Gene ID: | 55227 |
Gene Name: | leucine rich repeat containing 1 |
Gene Aliases: |
LANO, dJ523E19.1 |
Location: |
Chr.6:53794497-53924125 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC1 | NM_018214.4 | 1 | 163 | NP_060684.4 |
| XM_011514726.2 | 2 | 527 | XP_011513028.1 | |
| XM_017010997.1 | 1 | 455 | XP_016866486.1 | |
| AF332199.1 | 1 | 110 | AAK69623.1 | |
| AK021896.1 | 2 | 527 | BAB13929.1 | |
| AK292248.1 | ||||
| AU119761.1 | 2 | 527 | ||
| BC003193.1 | 1 | 163 | AAH03193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1183e214 | Chr.6:53765490 - 53799873 on Build GRCh38 | Loss |
|
| nsv1034955 | Chr.6:53582809 - 54066257 on Build GRCh38 | Gain |
|

Genomic Map