Assay Details
Target Gene Details
Entrez Gene ID: | 472 |
Gene Name: | ATM serine/threonine kinase |
Gene Aliases: |
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1 |
Location: |
Chr.11:108222484-108369102 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATM | NM_000051.3 | 1 | 126 | NP_000042.3 |
| XM_005271561.4 | 1 | 99 | XP_005271618.2 | |
| XM_011542842.2 | 1 | 100 | XP_011541144.1 | |
| XM_011542843.2 | 1 | 474 | XP_011541145.1 | |
| XM_011542844.2 | 1 | 98 | XP_011541146.1 | |
| XM_017017791.1 | 1 | 474 | XP_016873280.1 | |
| XM_017017792.1 | 1 | 474 | XP_016873281.1 | |
| X91196.1 | 1 | 474 | CAA62603.1 |
More Information
Set Membership: |
|

Genomic Map