Assay Details
Target Gene Details
Entrez Gene ID: | 221421 |
Gene Name: | radial spoke head 9 homolog |
Gene Aliases: |
C6orf206, CILD12, MRPS18AL1 |
Location: |
Chr.6:43645030-43671011 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSPH9 | NM_001193341.1 | NP_001180270.1 | ||
| NM_152732.4 | 3 | 515 | NP_689945.2 | |
| XM_005248901.3 | 3 | 488 | XP_005248958.1 | |
| XM_006715014.1 | 2 | 349 | XP_006715077.1 | |
| XM_017010426.1 | 3 | 488 | XP_016865915.1 | |
| AK055407.1 | BAB70918.1 | |||
| AK291399.1 | 3 | 488 | ||
| BC029519.1 | 3 | 472 | AAH29519.1 | |
| DA486918.1 | 2 | 350 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5281 | Chr.6:43611090 - 43655905 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map