Assay Details
Target Gene Details
Entrez Gene ID: | 128371 |
Gene Name: | olfactory receptor family 6 subfamily K member 6 |
Gene Aliases: |
OR1-21 |
Location: |
Chr.1:158754816-158755847 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6K6 | NM_001005184.1 | 1 | 423 | NP_001005184.1 |
| BC140743.1 | 1 | 519 |
Target Gene Details
Entrez Gene ID: | 128372 |
Gene Name: | olfactory receptor family 6 subfamily N member 1 |
Gene Aliases: |
OR1-22 |
Location: |
Chr.1:158747733-158840979 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6N1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764266 | Chr.1:158730942 - 158756090 on Build GRCh38 | Loss |
|
| dgv591n54 | Chr.1:158745498 - 158759556 on Build GRCh38 | Loss |
|
| dgv589n54 | Chr.1:158715246 - 158883163 on Build GRCh38 | Loss |
|
| dgv56e214 | Chr.1:158724216 - 158757728 on Build GRCh38 | Loss |
|
| nsv1002401 | Chr.1:158742837 - 159002241 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74122455] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map