Assay Details
Target Gene Details
Entrez Gene ID: | 6536 |
Gene Name: | solute carrier family 6 member 9 |
Gene Aliases: |
GLYT1 |
Location: |
Chr.1:43996483-44031492 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A9 | NM_001024845.2 | 14 | 2654 | NP_001020016.1 |
| NM_001261380.1 | 13 | 2601 | NP_001248309.1 | |
| NM_006934.3 | 13 | 2646 | NP_008865.2 | |
| NM_201649.3 | 14 | 2808 | NP_964012.2 | |
| NR_048548.1 | 13 | 2642 | ||
| XM_017002151.1 | 14 | 2562 | XP_016857640.1 | |
| XM_017002152.1 | 13 | 2536 | XP_016857641.1 | |
| XM_017002153.1 | 13 | 2307 | XP_016857642.1 | |
| AK304057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546142 | Chr.1:43990442 - 44000934 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map