Assay Details
Target Gene Details
Entrez Gene ID: | 100526794 |
Gene Name: | NT5C1B-RDH14 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:18554723-18589580 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5C1B-RDH14 | NM_001199103.1 | 9 | 1599 | NP_001186032.1 |
| NM_001199104.1 | 11 | 2048 | NP_001186033.1 | |
| AK057444.1 | 11 | 2030 | BAB71489.1 | |
| AK307653.1 | 9 | 1583 |
Target Gene Details
Entrez Gene ID: | 57665 |
Gene Name: | retinol dehydrogenase 14 (all-trans/9-cis/11-cis) |
Gene Aliases: |
PAN2, SDR7C4 |
Location: |
Chr.2:18554723-18560693 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDH14 | NM_020905.3 | 2 | 666 | NP_065956.1 |
| AF237952.1 | 2 | 608 | AAG12190.1 | |
| AK023625.1 | 1 | 993 | ||
| AK314994.1 | 2 | 626 | ||
| AY358511.1 | 2 | 609 | AAQ88875.1 | |
| BC009830.2 | 2 | 566 | AAH09830.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589966 | Chr.2:18553483 - 18559483 on Build GRCh38 | Loss |
|
| esv2751837 | Chr.2:18240747 - 18831848 on Build GRCh38 | Gain |
|
| nsv2624 | Chr.2:18527911 - 18562908 on Build GRCh38 | Insertion |
|
| nsv581114 | Chr.2:18200727 - 18615490 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map