Assay Details
Target Gene Details
Entrez Gene ID: | 80763 |
Gene Name: | spexin hormone |
Gene Aliases: |
C12orf39 |
Location: |
Chr.12:21526257-21532947 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPX | NM_030572.3 | NP_085049.1 | ||
| NR_135187.1 | ||||
| NR_135188.1 | ||||
| AK027273.1 | ||||
| AK075342.1 | ||||
| AK311147.1 | ||||
| AK311217.1 | ||||
| BC004336.1 | AAH04336.1 | |||
| DA071522.1 | ||||
| DA260291.1 | ||||
| DA380664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain |
|
| nsv1054220 | Chr.12:21459224 - 21539169 on Build GRCh38 | Gain |
|
| nsv557743 | Chr.12:21517476 - 21535966 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map