Assay Details
Target Gene Details
Entrez Gene ID: | 5024 |
Gene Name: | purinergic receptor P2X 3 |
Gene Aliases: |
P2X3 |
Location: |
Chr.11:57335943-57372399 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX3 | NM_002559.3 | NP_002550.2 | ||
| XM_011545069.2 | XP_011543371.1 | |||
| XM_011545070.2 | XP_011543372.1 | |||
| XM_011545071.2 | XP_011543373.1 | |||
| XM_011545072.2 | XP_011543374.1 | |||
| AB016608.1 | BAA76515.1 | |||
| BC069162.1 | AAH69162.1 | |||
| BC074792.2 | AAH74792.1 | |||
| BC074793.2 | AAH74793.1 | |||
| BC109287.2 | AAI09288.1 | |||
| DB462670.1 |
More Information
Set Membership: |
|

Genomic Map