Assay Details
Target Gene Details
Entrez Gene ID: | 11232 |
Gene Name: | DNA polymerase gamma 2, accessory subunit |
Gene Aliases: |
HP55, MTPOLB, PEOA4, POLB, POLG-BETA, POLGB |
Location: |
Chr.17:64477785-64497066 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLG2 | NM_007215.3 | 5 | 1147 | NP_009146.2 |
| XM_017024072.1 | 5 | 1100 | XP_016879561.1 | |
| AF142992.1 | 5 | 1130 | AAD50382.1 | |
| AF177201.1 | 5 | 1049 | AAD56640.1 | |
| AF184344.1 | 5 | 1130 | AAD56542.1 | |
| AK307801.1 | 5 | 1100 | ||
| BC000913.2 | 5 | 895 | ||
| BC009194.2 | 5 | 1117 | AAH09194.1 | |
| U94703.1 | 5 | 1131 | AAC51321.1 |
More Information
Set Membership: |
|

Genomic Map