Assay Details
Target Gene Details
Entrez Gene ID: | 1831 |
Gene Name: | TSC22 domain family member 3 |
Gene Aliases: |
DIP, DSIPI, GILZ, TSC-22R |
Location: |
Chr.X:107713221-107777329 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D3 | NM_001318468.1 | NP_001305397.1 | ||
| NM_001318470.1 | NP_001305399.1 | |||
| NM_004089.3 | NP_004080.2 | |||
| NM_198057.2 | NP_932174.1 | |||
| XM_005262099.1 | XP_005262156.1 | |||
| XM_005262100.1 | XP_005262157.1 | |||
| XM_005262102.1 | XP_005262159.1 | |||
| XM_005262103.3 | XP_005262160.1 | |||
| XM_011530884.1 | XP_011529186.1 | |||
| XM_017029335.1 | XP_016884824.1 | |||
| AB025432.1 | BAB18680.1 | |||
| AF153603.1 | AAD41085.1 | |||
| AF183393.1 | AAD56234.1 | |||
| AF228339.1 | AAG12456.1 | |||
| AK092645.1 | BAC03934.1 | |||
| AK092669.1 | ||||
| AK300535.1 | ||||
| AL110191.1 | CAB53669.1 | |||
| BC072446.1 | AAH72446.1 | |||
| BM047061.1 | ||||
| BX647854.1 | ||||
| CR933650.1 | CAI45951.1 |
More Information
Set Membership: |
|

Genomic Map