Assay Details
Target Gene Details
Entrez Gene ID: | 6596 |
Gene Name: | helicase like transcription factor |
Gene Aliases: |
HIP116, HIP116A, HLTF1, RNF80, SMARCA3, SNF2L3, ZBU1 |
Location: |
Chr.3:149030064-149086554 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HLTF | NM_001318934.1 | NP_001305863.1 | ||
| NM_001318935.1 | NP_001305864.1 | |||
| NM_003071.3 | NP_003062.2 | |||
| NM_139048.2 | NP_620636.1 | |||
| XM_011513091.1 | XP_011511393.1 | |||
| XM_011513092.1 | XP_011511394.1 | |||
| XM_011513093.1 | XP_011511395.1 | |||
| XM_017007078.1 | XP_016862567.1 | |||
| XM_017007079.1 | XP_016862568.1 | |||
| AB209052.1 | BAD92289.1 | |||
| AK291231.1 | ||||
| BC005260.1 | ||||
| BC015498.1 | ||||
| BC030976.1 | ||||
| BC044659.1 | AAH44659.1 | |||
| L34673.1 | AAA67436.1 |
Target Gene Details
Entrez Gene ID: | 100873945 |
Gene Name: | HLTF antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:149086332-149102823 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HLTF-AS1 | NR_046648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829753 | Chr.3:148954528 - 149120758 on Build GRCh38 | Gain |
|
| nsv528849 | Chr.3:149054135 - 149117654 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map