Assay Details
Target Gene Details
Entrez Gene ID: | 8382 |
Gene Name: | NME/NM23 family member 5 |
Gene Aliases: |
NM23-H5, NM23H5, RSPH23 |
Location: |
Chr.5:138115172-138139443 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME5 | NM_003551.2 | NP_003542.1 | ||
| BX490554.1 |
More Information
Set Membership: |
|

Genomic Map