Assay Details
Target Gene Details
Entrez Gene ID: | 4129 |
Gene Name: | monoamine oxidase B |
Gene Aliases: |
- |
Location: |
Chr.X:43766610-43882475 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAOB | NM_000898.4 | NP_000889.3 | ||
| XM_005272608.3 | XP_005272665.1 | |||
| XM_017029523.1 | XP_016885012.1 | |||
| XM_017029524.1 | XP_016885013.1 | |||
| AK294306.1 | ||||
| AK298942.1 | ||||
| AK312679.1 | ||||
| BC022494.1 | AAH22494.1 | |||
| M69177.1 | AAA59550.1 | |||
| S62734.1 | AAB27229.1 |
More Information
Set Membership: |
|

Genomic Map