Assay Details
Target Gene Details
Entrez Gene ID: | 54757 |
Gene Name: | family with sequence similarity 20 member A |
Gene Aliases: |
AI1G, AIGFS, FP2747 |
Location: |
Chr.17:68534561-68600954 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM20A | NM_001243746.1 | 12 | 2023 | NP_001230675.1 |
| NM_017565.3 | 11 | 1797 | NP_060035.2 | |
| NR_027751.1 | 11 | 1224 | ||
| XM_006721959.2 | 11 | 1169 | XP_006722022.1 | |
| AB545606.1 | 11 | 1797 | ||
| AF447872.1 | 5 | 1610 | AAQ04647.1 | |
| AK056789.1 | 12 | 1637 | BAB71285.1 | |
| AK298071.1 | ||||
| AL133105.1 | 9 | 906 | CAB61412.1 | |
| AY358197.1 | 12 | 1797 | AAQ88564.1 | |
| BC036222.1 | 11 | 1188 | AAH36222.1 | |
| BC136686.1 | 11 | 1686 | ||
| BC136689.1 | 12 | 1686 | ||
| CB530015.1 | 1 | 496 |
Target Gene Details
Entrez Gene ID: | 5573 |
Gene Name: | protein kinase cAMP-dependent type I regulatory subunit alpha |
Gene Aliases: |
ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1 |
Location: |
Chr.17:68413623-68551316 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAR1A | NM_001276290.1 | NP_001263219.1 | ||
| AW732104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833524 | Chr.17:68527889 - 68709053 on Build GRCh38 | Gain |
|
| nsv510723 | Chr.17:68533527 - 68561284 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map