Assay Details
Target Gene Details
Entrez Gene ID: | 84706 |
Gene Name: | glutamic--pyruvic transaminase 2 |
Gene Aliases: |
ALT2, GPT 2, MRT49 |
Location: |
Chr.16:46884380-46931297 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPT2 | NM_001142466.2 | NP_001135938.1 | ||
| NM_133443.3 | NP_597700.1 | |||
| AK094971.1 | BAC04465.1 | |||
| AY029173.2 | AAK31794.2 | |||
| BC062555.1 | AAH62555.1 | |||
| DA400677.1 | ||||
| HY117879.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
|
| nsv1063660 | Chr.16:46896711 - 46944880 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map