Assay Details
Target Gene Details
Entrez Gene ID: | 84316 |
Gene Name: | N(alpha)-acetyltransferase 38, NatC auxiliary subunit |
Gene Aliases: |
LSMD1, PFAAP2 |
Location: |
Chr.17:7856685-7885465 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA38 | NM_001320924.1 | NP_001307853.1 | ||
| NM_001320925.1 | NP_001307854.1 | |||
| NM_032356.4 | NP_115732.2 | |||
| XM_011524037.2 | XP_011522339.1 | |||
| XM_017025225.1 | XP_016880714.1 | |||
| XM_017025226.1 | XP_016880715.1 | |||
| AF530060.1 | AAQ09944.1 | |||
| BC006407.2 | AAH06407.1 | |||
| BC033861.1 | AAH33861.1 | |||
| BC051846.1 | AAH51846.1 | |||
| BC059944.1 | AAH59944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv510n67 | Chr.17:7852429 - 7911448 on Build GRCh38 | Gain |
|
| nsv833351 | Chr.17:7755691 - 7893863 on Build GRCh38 | Loss |
|
| nsv1070804 | Chr.17:7853581 - 7856982 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map