Assay Details
Target Gene Details
Entrez Gene ID: | 6431 |
Gene Name: | serine and arginine rich splicing factor 6 |
Gene Aliases: |
B52, HEL-S-91, SFRS6, SRP55 |
Location: |
Chr.20:43457864-43463605 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF6 | NM_006275.5 | 6 | 2899 | NP_006266.2 |
| NR_034009.1 | 7 | 3167 | ||
| AL110214.1 | 1 | 1427 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160678 | Chr.20:43432173 - 43469618 on Build GRCh38 | Deletion |
|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| nsv586029 | Chr.20:43303515 - 43493820 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map