Assay Details
Target Gene Details
| Entrez Gene ID: | 55370 | 
| Gene Name: | protein phosphatase 4 regulatory subunit 1 like (pseudogene) | 
| Gene Aliases: | C20orf192, PPP4R1P1, PRO1085 | 
| Location: | Chr.20:58232777-58309439 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 11 - Exon 12 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| PPP4R1L | NR_003505.3 | |||
| AK057583.1 | BAB71529.1 | |||
| CR994964.1 | ||||
| CX783537.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1055956 | Chr.20:58211957 - 58367256 on Build GRCh38 | Gain |  ANKRD60  PPP4R1L  RAB22A | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top