Assay Details
Target Gene Details
Entrez Gene ID: | 64106 |
Gene Name: | neuropeptide FF receptor 1 |
Gene Aliases: |
GPR147, NPFF1, NPFF1R1, OT7T022 |
Location: |
Chr.10:70254957-70283694 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPFFR1 | NM_022146.4 | 4 | 491 | NP_071429.1 |
| AB040104.1 | 3 | 473 | BAB17677.1 | |
| AF268898.1 | 3 | 473 | AAG41397.1 | |
| AF330055.1 | 3 | 473 | AAK94199.1 | |
| BC131580.1 | 3 | 473 | ||
| EU432128.1 | 3 | 473 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv1041698 | Chr.10:70244698 - 70306282 on Build GRCh38 | Gain |
|
| nsv831908 | Chr.10:70210726 - 70389945 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map