Assay Details
Target Gene Details
Entrez Gene ID: | 23647 |
Gene Name: | ADP ribosylation factor interacting protein 2 |
Gene Aliases: |
POR1 |
Location: |
Chr.11:6474683-6481479 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARFIP2 | NM_001242854.1 | NP_001229783.1 | ||
| NM_001242855.1 | NP_001229784.1 | |||
| NM_001242856.1 | NP_001229785.1 | |||
| NM_012402.3 | NP_036534.1 | |||
| XM_005252840.4 | XP_005252897.1 | |||
| AK026390.1 | ||||
| AK300855.1 | ||||
| AK301856.1 | ||||
| AK301972.1 | ||||
| AK304512.1 | ||||
| AK310267.1 | ||||
| AK311004.1 | ||||
| BC000392.2 | AAH00392.1 | |||
| U52522.1 | AAA97924.1 | |||
| X97567.1 | CAA66179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625266 | Chr.11:6460034 - 6490367 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map