Assay Details
Target Gene Details
Entrez Gene ID: | 23137 |
Gene Name: | structural maintenance of chromosomes 5 |
Gene Aliases: |
SMC5L1 |
Location: |
Chr.9:70258962-70354873 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMC5 | NM_015110.3 | 1 | 187 | NP_055925.2 |
| XM_005251837.1 | 1 | 187 | XP_005251894.1 | |
| XM_005251838.1 | 1 | 187 | XP_005251895.1 | |
| XM_005251839.1 | 1 | 187 | XP_005251896.1 | |
| XM_017014507.1 | 1 | 187 | XP_016869996.1 | |
| XM_017014508.1 | 1 | 187 | XP_016869997.1 | |
| XM_017014509.1 | 1 | 187 | XP_016869998.1 | |
| AB011166.2 | 1 | 128 | ||
| AJ310550.1 | 1 | 70 | CAC39247.1 | |
| AK055825.1 | 1 | 100 | ||
| AK304133.1 | 1 | 187 | ||
| BC017666.1 | 1 | 146 | ||
| BC038225.1 | 1 | 170 | AAH38225.1 | |
| DB220819.1 | 1 | 187 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6555 | Chr.9:70225054 - 70259811 on Build GRCh38 | Insertion |
|
| nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain |
|
| nsv831615 | Chr.9:70237789 - 70404776 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map