Assay Details
Target Gene Details
Entrez Gene ID: | 151126 |
Gene Name: | zinc finger protein 385B |
Gene Aliases: |
ZNF533 |
Location: |
Chr.2:179441982-179864550 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF385B | NM_001113397.1 | 8 | 1664 | NP_001106868.1 |
NM_001113398.1 | 8 | 1524 | NP_001106869.1 | |
NM_001282725.1 | 10 | 1618 | NP_001269654.1 | |
NM_152520.4 | 10 | 2191 | NP_689733.3 | |
NR_104234.1 | ||||
XM_011510713.2 | 12 | 5332 | XP_011509015.1 | |
XM_011510714.2 | 11 | 5282 | XP_011509016.1 | |
XM_011510715.2 | 11 | 5235 | XP_011509017.1 | |
XM_011510716.2 | 11 | 2443 | XP_011509018.1 | |
XM_011510717.2 | 11 | 2440 | XP_011509019.1 | |
XM_011510719.2 | 11 | 1703 | XP_011509021.1 | |
XM_011510720.2 | 10 | 1563 | XP_011509022.1 | |
XM_011510721.2 | 9 | 3856 | XP_011509023.1 | |
XM_017003435.1 | 10 | 1872 | XP_016858924.1 | |
XM_017003436.1 | 10 | 2350 | XP_016858925.1 | |
XM_017003437.1 | 9 | 1507 | XP_016858926.1 | |
XM_017003438.1 | 8 | 6990 | XP_016858927.1 | |
XM_017003439.1 | 8 | 1498 | XP_016858928.1 | |
XM_017003440.1 | 9 | 5027 | XP_016858929.1 | |
AK057999.1 | BAB71629.1 | |||
AK096822.1 | 8 | 1664 | BAC04870.1 | |
BC038422.1 | 10 | 1616 | AAH38422.1 | |
BC048123.1 | AAH48123.1 | |||
BC092423.1 | 10 | 2026 | AAH92423.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
