Assay Details
Target Gene Details
Entrez Gene ID: | 51279 |
Gene Name: | complement C1r subcomponent like |
Gene Aliases: |
C1RL1, C1RLP, C1r-LP, CLSPa |
Location: |
Chr.12:7093662-7109278 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1RL | NM_001297640.1 | 5 | 1365 | NP_001284569.1 |
| NM_001297642.1 | 6 | 1538 | NP_001284571.1 | |
| NM_016546.3 | 6 | 1491 | NP_057630.2 | |
| XM_017019397.1 | 6 | 1538 | XP_016874886.1 | |
| XM_017019398.1 | 5 | 1412 | XP_016874887.1 | |
| XM_017019400.1 | XP_016874889.1 | |||
| XM_017019402.1 | XP_016874891.1 | |||
| AF178985.1 | 6 | 1410 | AAF44349.1 | |
| AK024084.1 | BAB14819.1 | |||
| AK222802.1 | 6 | 1418 | BAD96522.1 | |
| AK314299.1 | 6 | 1498 |
More Information
Set Membership: |
|

Genomic Map