Assay Details
Target Gene Details
Entrez Gene ID: | 23239 |
Gene Name: | PH domain and leucine rich repeat protein phosphatase 1 |
Gene Aliases: |
PHLPP, PLEKHE1, PPM3A, SCOP |
Location: |
Chr.18:62715439-62980443 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHLPP1 | NM_194449.3 | 1 | 1711 | NP_919431.2 |
| XM_011525886.1 | 1 | 1705 | XP_011524188.1 | |
| AB011178.2 | 1 | 424 | BAA25532.2 | |
| AK001924.1 | BAA91980.1 | |||
| AL079409.1 | 2 | 108 | ||
| BC014927.2 | ||||
| BC126277.1 |
More Information
Set Membership: |
|

Genomic Map