Assay Details
Target Gene Details
Entrez Gene ID: | 140733 |
Gene Name: | MACRO domain containing 2 |
Gene Aliases: |
C20orf133 |
Location: |
Chr.20:13995476-16053197 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MACROD2 | NM_080676.5 | 1 | 125 | NP_542407.2 |
| XM_017027677.1 | 1 | 149 | XP_016883166.1 | |
| AK131348.1 | 1 | 256 | BAD18504.1 | |
| BX281637.1 | ||||
| DB125529.1 | 1 | 256 |
More Information
Set Membership: |
|

Genomic Map