Assay Details
Target Gene Details
Entrez Gene ID: | 497661 |
Gene Name: | chromosome 18 open reading frame 32 |
Gene Aliases: |
- |
Location: |
Chr.18:49481178-49487274 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C18orf32 | NM_001035005.3 | NP_001030177.1 | ||
| NM_001199346.1 | NP_001186275.1 | |||
| AB097012.1 | BAC77365.1 | |||
| AK027111.1 | ||||
| AK291882.1 | ||||
| BC022357.1 | AAH22357.1 | |||
| BC093004.1 | AAH93004.1 | |||
| BF210402.1 |
Target Gene Details
Entrez Gene ID: | 100526842 |
Gene Name: | RPL17-C18orf32 readthrough |
Gene Aliases: |
- |
Location: |
Chr.18:49481178-49492565 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL17-C18orf32 | NM_001199355.1 | NP_001186284.1 | ||
| NM_001199356.1 | NP_001186285.1 | |||
| BI461805.1 | ||||
| BX393840.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv978629 | Chr.18:49486706 - 49491456 on Build GRCh38 | Duplication |
|
| nsv2294 | Chr.18:49470841 - 49492546 on Build GRCh38 | Deletion |
|
| nsv1065279 | Chr.18:49468523 - 49512116 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map